Would you like to log in again?
Babcock Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
36-1662 36-1677 36-1692 22-6155
89025-378EA 180.44 USD InStock
89025-378 94002-748 10195-726 10192-444
Babcock Forceps, OR Grade, Sklar
Forceps
Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue. They are frequently used with intestinal and laparotomy procedures.
Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface
Censitrac Ready OR Grade Babcock Forceps are industry standard and constructed of stainless steel.
The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty