Would you like to log in again?
Ferris-Smith Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
52-3970
94002-752EA 246.21 USD InStock
94002-752
Ferris-Smith Tissue Forceps, OR Grade, Sklar
Forceps
The Ferris Smith Tissue Forceps are heavyweight forceps used with fibrous tissue, commonly in colorectal procedures
These thumb forceps feature a wide section of the handle for greater control handling firm tissue. They also have slightly wider tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty