Would you like to log in again?
Master Lock® Grip Tight Circuit Breaker Lockout, NMC
Supplier: NMC (National Marker Company)
491B 493B
94002-858EA 29.5 USD InStock
94002-858 94002-860
Master Lock® Grip Tight Circuit Breaker Lockout, NMC
Security Locks
Grip tight with simple thumb turn and clamping handle
Accommodates virtually all styles of breaker toggles. Designed to permit side-by-side breaker lockout. Suitable for harsh environments. Powder coated steel and reinforced polymer allows for operation in harsh environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty