Would you like to log in again?
Precoiled Pipe Markers- Sanitary Sewer W/G, National Marker
Supplier: National Marker
C4094 D4094 A4094 F4094 B4094 E4094 G4094 H4094
94003-468EA 16.1 USD InStock
94003-468 94013-926 94009-630 94004-242 94012-730 94005-700 94015-252 94014-472
Precoiled Pipe Markers- Sanitary Sewer W/G, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty