Would you like to log in again?
Precoiled Pipe Markers 'Plant Air', Black/Yellow, National Marker
Supplier: National Marker
F4081 D4081 C4081 A4081 E4081 H4081 G4081 B4081
94015-226EA 87.99 USD InStock
94015-226 94013-900 94009-596 94014-446 94005-668 94003-442 94012-704 94004-216
Precoiled Pipe Markers 'Plant Air', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty