Would you like to log in again?
Precoiled Pipe Markers- Supply B/Y, National Marker
Supplier: National Marker
F4102 B4102 E4102 H4102 G4102 C4102 A4102 D4102
94015-268EA 87.99 USD InStock
94015-268 94009-664 94013-942 94005-722 94012-746 94003-484 94014-488 94004-258
Precoiled Pipe Markers- Supply B/Y, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty