Would you like to log in again?
NMC Large Circuit Breaker Lockout
Supplier: NMC (National Marker Company)
BL05
94004-310EA 87.01 USD InStock
94004-310
NMC Large Circuit Breaker Lockout
Security Locks
Lockouts out circuit breakers with single toggles up to 2.03cm (.8") thick and 7.62cm (3") wide
Easy installation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty