Would you like to log in again?
Precoiled Pipe Markers- 'Compressed Air' W/B, National Marker
Supplier: National Marker
C4022 G4022 F4022 B4022 D4022 H4022 E4022 A4022
94014-328EA 64.41 USD InStock
94014-328 94005-494 94003-324 94004-098 94012-586 94015-108 94013-782 94009-406
Precoiled Pipe Markers- 'Compressed Air' W/B, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty