Would you like to log in again?
Precoiled Pipe Markers- 'Medical Air' W/B, National Marker
Supplier: National Marker
D4071 A4071 C4071 F4071 G4071 B4071 H4071 E4071
94012-684EA 46.11 USD InStock
94012-684 94004-196 94015-206 94014-426 94005-636 94013-880 94009-568 94003-422
Precoiled Pipe Markers- 'Medical Air' W/B, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty