Alert employees and guests to smoking policies within your facility.
- Ideal for indoor and outdoor labeling
- Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
- .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
- .004 PS vinyl material has pressure sensitive adhesive backing
- .095 fiber glass material is rigid, shatter-proof and dent resistant
- .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
No smoking signs indicate where flammable materials present a fire risk. Many states now have no smoking laws, post these no smoking signs around your facility as visual reminders to No Smoking policies.