Would you like to log in again?
Personal Protection (PPE), OSHA Caution Signs, National Marker
Supplier: National Marker
C506RB C502RB C600PB C518AB C499RB C514AB C517RB C600RB C602RB C520PB C520AB C601PB C588PB C507AB C518PB C600AB C487RB C603RB C486PB C513RB C598RC C513AB C507RB C519AB C599PB C507PB C521PB C484PB C588AB C506AB C484RB C603PB C521RB C519RB C499PB C516PB C632PB C518RB C487PB C598AD C602PB C501AB C515AB C485AB C504PB C506PB C598RD C514RB C503RB C510AP C502PB C503PB C486RB C599RB C603AB C632RB C598AC C501PB C485RB C484AB C602AB C588RB C503AB C599AB C499AB C598AB C517PB C516RB C519PB C515PB C601AB C504AB C601RB C515RB C517AB C520RB C598PB C485PB C598RB C501RB C521AB C516AB C504RB C632AB C486AB C502AB C487AB C514PB C513PB
94006-480EA 29.4 USD InStock
94006-480 94006-484 94006-440 94006-482 94007-046 94007-048 94007-050 94007-052 94007-054 94006-478 94006-434 94006-950 94006-476 94006-432 94006-398 94006-438 94006-436 94006-250 94006-370 94006-452 94006-254 94006-374 94006-252 94006-450 94006-372 94007-238 94007-042 94007-240 94007-044 94007-242 94006-368 94006-400 94006-444 94006-246 94006-366 94006-486 94006-442 94006-448 94006-248 94006-446 94006-382 94006-260 94006-380 94006-386 94006-462 94006-264 94006-340 94006-384 94006-460 94006-262 94007-024 94007-026 94007-028 94007-030 94007-032 94006-456 94006-258 94006-378 94006-454 94006-256 94006-376 94006-338 94006-458 94006-414 94006-336 94006-392 94006-390 94006-430 94006-396 94006-394 94007-056 94007-058 94007-014 94007-016 94007-018 94006-948 94007-060 94006-946 94007-062 94006-428 94007-064 94007-020 94007-022 94006-268 94006-388 94006-464 94006-266 94006-426 94006-424
Personal Protection (PPE), OSHA Caution Signs, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads, "Caution".
Certifications: Comply with OSHA 29 CFR 1910.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty