Would you like to log in again?
Machine Signs, 0.040 Alum, National Marker
Supplier: National Marker
C454AB C539AB D149AB DGA48AB SF154AB N140AB C596AB C534AB C538AB N124AB D477AB C15AB W419AB C81AB C467AB SPD87AB D213AB SF167AB DGA46AB C618AB D567AB D600AB W403AB C171AB SPD365AB DGA55AB C623AB C640AB ESC700AB C79AB C610AB D509AB D505AB ESD466AB W450AB D105AB C532AB D621AB C535AB W451AB SPC700AB D466AB D431AB D449AB D507AB N267AB D599AB C619AB D261AB D608AB C457AB D455AB D87AB C595AB W459AB D479AB C622AB C594AB D305AB C621AB ESW501AB SPW501AB D618AB C399AB D508AB D465AB C461AB D510AB C497AB C353AB C463AB W464AB ESD365AB D566AB C536AB D564AB D565AB C613AB
94010-302EA 51.84 USD InStock
94010-302 94021-678 94006-084 94021-512 94023-362 94013-712 94023-126 94006-560 94006-120 94021-594 94007-562 94008-492 94011-866 94011-108 94009-854 94007-172 94008-202 94011-874 94008-682 94011-192 94006-994 94011-270 94021-948 94010-178 94010-018 94010-498 94010-894 94020-980 94007-552 94007-158 94010-290 94005-134 94008-914 94007-286 94007-120 94006-566 94006-324 94004-812 94004-856 94017-656 94010-888 94006-144 94021-058 94010-166 94006-066 94010-486 94009-076 94010-882 94006-582 94011-922 94007-102 94007-150 94013-464 94006-574 94011-252 94010-516 94013-262 94023-232 94010-078 94023-312 94010-510 94007-178 94005-422 94023-392 94018-230 94011-144 94007-184 94006-588 94017-636 94023-306 94013-412 94006-982 94006-548 94011-460 94006-988 94010-504 94006-108 94010-902
Machine Signs, 0.040 Alum, National Marker
Signs
Signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Made of 0.040 aluminum
  • Weatherproof and non-corrosive, making it ideal for indoor and outdoor use
  • Baked-on enamel on both sides, allowing it to be printed on both sides
Avoid accidents and keep employees safe by clearly identifying hazardous equipment situations. This can include heavy equipment, moving machine parts, powered vehicles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty