Would you like to log in again?
Admittance and Security Danger Signs, Do Not Enter, National Marker
Supplier: National Marker
SPD81PB SPD104PB SPD104AB ESD104PB D104AD D499PB D499RB D500PB D104RC SPD104RB D490AB SPD81AB ESD104RB D428PB D500RB D499AB D428AB D104RD D104AC D478RB D500AB D478AB ESD104AB D490RB D478PB SPD81RB D490PB
94021-536EA 20.3 USD InStock
94021-536 94021-534 94021-676 94021-674 94010-300 94021-672 94010-460 94009-834 94009-832 94008-200 94010-456 94010-458 94010-430 94010-298 94010-432 94008-186 94010-372 94008-184 94010-296 94010-374 94010-370 94013-294 94013-292 94013-290 94008-198 94010-428 94021-538
Admittance and Security Danger Signs, Do Not Enter, National Marker
Signs
Alert visitors and guest of security policies and direct them towards appropriate entrances
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Each sign header reads "Danger".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty