Would you like to log in again?
Chemical OSHA Danger Signs, Formaldehyde, National Marker
Supplier: National Marker
D537AB D170R D537PB D537RB
94010-698EA 29.06 USD InStock
94010-698 94010-694 94010-696 94008-582
Chemical OSHA Danger Signs, Formaldehyde, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .095 fiber glass material is rigid, shatter-proof and dent resistant
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads "Danger".
Certifications: Comply with OSHA 29 CFR 1910.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty