Would you like to log in again?
Voltage OSHA Danger Signs, Fiberglass, National Marker
Supplier: National Marker
D101EB D290EB D421EB D100EB D473EB D139EB D289EB D549EB D443EB D325EB D49EB D444EB
94009-754EA 59.41 USD InStock
94009-754 94009-982 94010-782 94009-176 94010-266 94009-970 94009-002 94008-164 94008-172 94010-440 89185-870 94008-996
Voltage OSHA Danger Signs, Fiberglass, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Made of .095 fiberglass
  • Rigid, shatter-proof and dent resistant
  • Resistant to high traffic, severe outdoor weather conditions and weak alkalis and acids
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads, "Danger."
Certifications: Comply with OSHA 29 CFR 1910.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty