Would you like to log in again?
Precoiled Pipe Markers- 'Propane Gas', Black/Yellow, National Marker
Supplier: National Marker
C4086 H4086 G4086 E4086 A4086 F4086 D4086 B4086
94015-236EA 87.99 USD InStock
94015-236 94014-456 94013-910 94005-678 94012-714 94009-606 94003-452 94004-226
Precoiled Pipe Markers- 'Propane Gas', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty