Would you like to log in again?
Precoiled Pipe Markers- Steam Return B/Y, National Marker
Supplier: National Marker
C4098 A4098 H4098 B4098 E4098 G4098 F4098 D4098
94005-708EA 41.82 USD InStock
94005-708 94013-934 94014-480 94004-250 94015-260 94009-638 94012-738 94003-476
Precoiled Pipe Markers- Steam Return B/Y, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty