Would you like to log in again?
Precoiled Pipe Markers 'High Pressure Air', Black/Yellow, National Marker
Supplier: National Marker
H4057 D4057 F4057 G4057 A4057 B4057 C4057 E4057
94009-522EA 42.82 USD InStock
94009-522 94004-168 94005-594 94003-394 94012-656 94013-852 94014-398 94015-178
Precoiled Pipe Markers 'High Pressure Air', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty