Would you like to log in again?
Precoiled Pipe Markers- 'High Pressure Condensate', Black/Yellow, National Marker
Supplier: National Marker
E4058 F4058 A4058 D4058 B4058 H4058 C4058 G4058
94005-596EA 41.82 USD InStock
94005-596 94013-854 94015-180 94004-170 94012-658 94003-396 94014-400 94009-524
Precoiled Pipe Markers- 'High Pressure Condensate', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty