Would you like to log in again?
Precoiled Pipe Markers 'Hot Water', Black/Yellow, National Marker
Supplier: National Marker
D4061 F4061 C4061 H4061 B4061 E4061 G4061 A4061
94003-402EA 16.08 USD InStock
94003-402 94014-406 94004-176 94009-540 94005-606 94012-664 94015-186 94013-860
Precoiled Pipe Markers 'Hot Water', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty