Would you like to log in again?
Precoiled Pipe Markers 'Oxygen', White/Green, National Marker
Supplier: National Marker
E4080 B4080 H4080 F4080 G4080 C4080 D4080 A4080
94015-224EA 87.99 USD InStock
94015-224 94013-898 94009-594 94012-702 94003-440 94014-444 94004-214 94005-666
Precoiled Pipe Markers 'Oxygen', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty