Would you like to log in again?
Precoiled Pipe Markers 'Domestic Cold Water Supply', White/Green, National Marker
Supplier: National Marker
G4036 A4036 C4036 H4036 B4036 D4036 E4036 F4036
94015-136EA 87.99 USD InStock
94015-136 94009-454 94013-810 94003-352 94012-614 94014-356 94005-532 94004-126
Precoiled Pipe Markers 'Domestic Cold Water Supply', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty