Would you like to log in again?
Precoiled Pipe Markers- 'Direction Arrow', Black/Yellow, National Marker
Supplier: National Marker
E4115 B4115 D4115 C4115 H4115 G4115 A4115
94009-698EA 42.82 USD InStock
94009-698 94012-772 94004-284 94014-514 94015-294 94005-754 94003-510
Precoiled Pipe Markers- 'Direction Arrow', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty