Would you like to log in again?
MasterLock® Snap-On Advantage Hasp, NMC
Supplier: NMC (National Marker Company)
SL115
94021-302EA 21.06 USD InStock
94021-302
MasterLock® Snap-On Advantage Hasp, NMC
Security Locks
Lock out your equipment and machinery in a snap
Features a spring loaded jaw end. Padlock inserted into the holes prevents the jaws from opening. Accommodates up to 5 padlocks. Hasp made from durable anodized aluminum and steel rivet construction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty