Would you like to log in again?
NMC Valve Wheel Lockouts
Supplier: NMC (National Marker Company)
VL04R VL09R VL02R VL13R VL06R
94022-908EA 114.21 USD InStock
94022-908 94022-904 94022-906 94022-910 94022-912
NMC Valve Wheel Lockouts
Security Locks
Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter
Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty