Would you like to log in again?
VISI SIGNS™, National Marker
Supplier: National Marker
VS2R VS51W VS11R VS15W VS16R VS45W VS10W VS9R VS33W VS46W VS52W VS28W VS40W VS44W VS27W VS16W VS31W VS19W VS15R VS4W VS11W VS5W VS3R VS49W VS38W VS42W VS20R VS9W VS41W VS50W VS21W VS13R VS2W VS19R VS4R VS5R VS1W VS20W VS13W VS3W VS10R
94023-002EA 86.7 USD InStock
94023-002 94023-000 94023-006 94023-004 94022-936 94022-938 94022-976 94022-954 94022-998 94022-978 94022-934 94022-956 94022-940 94022-984 94022-942 94022-964 94022-986 94022-980 94022-982 94023-010 89185-998 89185-978 94022-926 94022-948 89185-994 94022-928 94022-966 94022-944 94022-988 94022-922 94022-994 94022-972 94022-974 94022-996 94022-952 94022-990 94022-970 94022-992 89186-002 94023-008 89186-000
VISI SIGNS™, National Marker
Signs
An alternative to flat signs, Visi-Signs™ stand out from the wall, allowing for viewing from multiple directions
Made from durable 3mm thick Ultra White PVC . Projects from wall at 45⁰ angle. Features pre-drilled mounting holes for fast and easy installation. Excellent fire properties with a UL 94 V-0 flammability rating.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty