Would you like to log in again?
Tweezers, Straight Delrin Soft Replaceable Tips, Excelta Corp®
Supplier: Excelta
159D-RTWX 159D-RTWX 159B-RTWX 159C-RTWX
10066-508EA 10.22 USD InStock
10066-508 76243-388 94024-106 94024-116
Tweezers, Straight Delrin Soft Replaceable Tips, Excelta Corp®
Forceps
These soft replaceable tip tweezers feature Acetal tips which are electrically insulative and resistant to temperatures up to 185 °F.
The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). Excellent for cleanroom applications and for avoiding the marring or scratching of items being handled.
All replacement tips come complete with 2 tips and 4 screws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty