Would you like to log in again?
Tweezers, Carbofib Soft Replaceable Tip, Excelta Corp®
Supplier: Excelta
159-RT 159D-RT 159F-RT 159-RT 159D-RT 159G-RT 159D-RTN 159A-RT 159A-RT 759D-RT 159B-RT 159-RTN 759-RT 159-RTN 159B-RT 159C-RT 159-RT-ET 759-RTN 159E-RT
63046-724EA 62.46 USD InStock
63046-724 76471-534 76471-536 75880-634 76252-144 75880-844 75880-636 10066-510 63043-714 63041-264 76471-540 10066-512 94024-118 16003-848 76240-114 89411-864 76238-648 76238-654 76238-644
Tweezers, Carbofib Soft Replaceable Tip, Excelta Corp®
Forceps
Carbon Fiber tipped tweezers prevent nicking and marring of delicate items while maintaining high rigidity and tensil strength.
  • Replaceable tips
  • Available in a multitude of tip sizes
  • Temperature resistant to 249 °F
  • Conductive
  • Will not scratch or mar parts
  • Excellent tensile and flexural strength
These soft replaceable tip tweezer boasts Carbofib™ tips. The Carbofib™ tips are a polymer based material with 30% carbon fibers added (conductive 10² ohms/sq). Carbofib™ has very high rigidity, fatigue and creep resistance. The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). They are excellent for handling delicate items.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty