Would you like to log in again?
Four Star™ Tweezer/Forceps, Mirror Polished Tip, EXCELTA®
Supplier: Excelta
5-SA-MP M-2-SA-MP 51-SA-MP 1-SA-MP 2A-SA-MP 3C-SA-MP 7-SA-MP 5A-SA-MP
94024-384EA 157.18 USD InStock
94024-384 94024-152 89411-870 94024-302 94024-304 94024-216 94024-282 94024-260
Four Star™ Tweezer/Forceps, Mirror Polished Tip, EXCELTA®
Forceps
Our Four Star series with mirror polished tips are designed for use under a scope
These forceps are made from Neverust Stainless Steel that is Anti-magnetic.
The extreme polishing of the inside edges prevents any possible nicking or scratching.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty