Would you like to log in again?
Brushes, Cleanroom Safe, Excelta Corp®
Supplier: Excelta
213A-N 213D-N 213C-N 213B-N
94024-188EA 27.71 USD InStock
94024-188 94024-190 94024-192 94024-194
Brushes, Cleanroom Safe, Excelta Corp®
Brushes
The 'Series 213' cleanroom safe brush has stainless steel handles and new Statigo9 fiber bristles.
  • Designed for cleaning in tight areas
  • Stainless steel handle, corrosion resistant
The bristles are mechanically clamped into the handle for maximum fiber retention. The fibers are similar to nylon; they have a surface resistivity of 10⁸ ohms/sq.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty