Would you like to log in again?
Pliers, Bent Nose, Excelta
Supplier: Excelta
2829 2829D 2829L 2629 2914D 2629 2904 2829-CR 2829LD 2904 2914
63041-788EA 58.51 USD InStock
63041-788 63041-786 94024-210 75880-932 76240-950 76471-590 76471-592 94024-204 63041-706 76244-928 76471-620
Pliers, Bent Nose, Excelta
Pliers
Made of Stainless Steel, these bent nose pliers offer excellent visibility and accessibiity.
  • Bent nose affords greater accessibility, visibility
  • Ergonomic handle, dual leaf springs
  • Stainless steel construction, corrosion resistant
  • Different sizes depending on application
  • Most available with serrated grips
Bent nose pliers are made of AISI 440 stainless steel and include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application. Bent nose pliers offer greater accessability and more visibility when work in confined areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty