Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Probes, Very Fine Tip (0.001"), Stainless Steel, Excelta Corp®
Supplier: Excelta
332B 335B 335B 335A 332A 332D 332E 332C
76243-468EA 23.68 USD InStock
76243-468 94024-220 94024-222 94024-224 89411-732 94024-226 94024-228 89411-730
Probes, Very Fine Tip (0.001"), Stainless Steel, Excelta Corp®
Dissection Needles
Smallest tip made - great for magnification and very fine work.
  • Extreme fine tips for extreme fine work
  • Available in short or long handle
  • Corrosion resistant
These stainless steel probes have a 1 mil tip. The 335 Series Mini-Probes are 4.75" long. The 332 series are 6.5" long. They feature the smallest, finest tips available on any probe and are ideal for microscopy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty