Would you like to log in again?
Pliers, Tubing Expander, Reverse Action, Excelta
Supplier: Excelta
Two Star™
379C 379A 379B
94024-240EA 58 USD InStock
94024-240 94024-242 94024-244
Pliers, Tubing Expander, Reverse Action, Excelta
Pliers
These stainless steel reverse action pliers have been specifically designed to expand the end of flexible tubing for inserting into plastic or metal fittings and connections. The tools are 5" long and expands tubing to 3 different size openings. They are spring loaded for ease of use.
  • Expands tubing of various sizes
  • Reverse action for ease of use
  • Cushioned grips/double leaf springs
Designed to expand the end of flexible tubing for inserting into plastic or metal fittings and connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty