Would you like to log in again?
Scribes, Diamond Tipped, Excelta Corp®
Supplier: Excelta
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
475A 475B
94024-264EA 69.24 USD InStock
94024-264 94024-266
Scribes, Diamond Tipped, Excelta Corp®
Pens Stylus and Scribers
Diamond coated tips are very strong.
  • Light weight aluminum handle
  • Ideal for scraping
These diamond tipped scribes with aluminum handles and 20 mil fine tip are ideal for scraping.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty