Would you like to log in again?
Wafer Tweezers, Excelta Corp®
Supplier: Excelta
891-SA 890L-SA
63042-998EA 336.36 USD InStock
63042-998 94024-390
Wafer Tweezers, Excelta Corp®
Forceps
These tweezers are specifically designed handling 6" wafers.
  • For scratch free holding of 8" wafers
  • Cleanroom safe, anti-mag. Stainless steel
  • Various configurations for various applications
The wafer tweezers for 8" wafers are made of stainless steel and are anti-magnetic. They come in a variety of configurations. The 'B' version features a 20° bend in the body to improve handling. The overall length is 4.75".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty