Would you like to log in again?
SatPax® 1200-R Pre-Wetted Cleanroom Wipers, Berkshire
Supplier: Berkshire
SPX120R00112
94024-546CS 800.18 USD InStock
94024-546
SatPax® 1200-R Pre-Wetted Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® 1200-R combines SuperPolx® 1200 cleanroom laundered wipers composed of continuous filament polyester knit fabric with a high saturation level of 70% IPA and 30% DI water.
  • Very low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
  • Smooth and durable with good wet strength
  • Increases cleaning protocol consistency
This pre-wetted format provides a solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper. A high solution saturation level of 60% is suitable for regulated applications or applications requiring higher solution levels delivered to the surface.
Laundered and packaged in ISO class 4 (class 10) cleanroom. Available sterile for aseptic processing. Recommended for use in ISO class 4 (class 10) cleanroom applications and above. Reduce storage requirements and usage quantities. Eliminate squirt bottles and uncontrolled saturation levels Reduces VOC (Volatile Organic Compound) emissions. Available in a flat pack, or J-fold configurations.
Designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.
Certifications: Complies with RoHS and REACH requirements.
Ordering information: 30 sheets per pack, 12 packs per case (double packed).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty