Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yeast Extract, Powder, Certified, HiMedia Laboratories

Supplier: HiMedia Laboratories
CR027-25KG CR027-500G CR027-25KG CR027-500G
95041-228EA 192.83 USD InStock
CA95041-228 95041-228 95000-018 CA95000-018
Yeast Extract, Powder, Certified, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
It is a certified grade manufactured under controlled condition by autolysis of Saccharomyces cerevesiae

  • Light yellow to brownish yellow colored
  • Homogenous, free flowing powder with characteristic odour but not putrescent
  • Soluble in distilled/purified water
  • Insoluble in alcohol and ether
  • Reaction of 2% w/v aqueous solution at 25 ºC

It is rich in vitamins, especially those belonging to B-complex and used particularly for cultivation of microorganisms encountered in milk or other dairy products.

Caution: Store below 30 °C.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty