Would you like to log in again?
Proteose Peptone A, HiMedia
Supplier: HiMedia Laboratories
RM6394-500G RM6394-10KG RM6394-2.5KG RM6394-2.5KG RM6394-10KG RM6394-25KG RM6394-5KG RM6394-5KG RM6394-500G RM6394-25KG
95029-586EA 209.4 USD InStock
CA95007-737 95029-586 CA95007-735 95007-737 CA95029-586 95008-100 CA95008-100 95007-735 CA95007-733 95007-733
Proteose Peptone A, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Proteose Peptone A is recommended for cultivation of fastidious pathogens and particularly for bulk production of antibiotics, enzymes, veterinary preparations and bacterial toxins especially diphtheria toxin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty