Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1 N

Supplier: HiMedia
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

TCL002-20ML TCL002-100ML TCL002-20ML TCL002-10X20ML
95016-356PK 143.11 USD InStock
95016-356 CA76263-430 CA76203-568 76203-568
Sodium hydroxide 1 N
Sodium hydroxide
Sodium hydroxide (1 N) solution prepared in cell culture grade water. It is used to buffer cell culture medium as per application.

  • Sterile filtered
  • Appearance: Colorless, clear liquid
  • pH: 12.00 to14.00


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty