Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®

Supplier: HiMedia Laboratories

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TS1006-20L TS1006-1L TS1006-5L TS1006-1L TS1006-5L TS1006-20L
95016-730EA 204.29 USD InStock
CA95016-730 95016-730 95016-726 95016-728 CA95016-728 CA95016-726
Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Buffers Cell Culture Buffers
Dulbecco's phosphate buffered saline is used for tissue disaggregation and monolayer dispersal.The presence of calcium and magnesium ions hinder the trypsin activity.

  • Off-white to creamish white
  • pH: 7.20 to 7.80
  • Osmolality without sodium bicarbonate: 275.00 to 315.00 mOsm
  • Endotoxin content: NMT 1 EU/ml

The solution is used to maintain the medium within physiological pH range and intracellular and extra cellular osmotic balance. Modified with a carbohydrate, such as glucose serves as an energy source for cell metabolism.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty