Would you like to log in again?
Sephadex™ G-25 Gel Filtration Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sephadex™
17-0034-01 17-0032-01 17-0032-01 17-0033-01 17-0031-01 17-0034-02 17-0031-01 17-0033-02 17-0032-02 17-0033-02 17-0033-01 17-0034-02 17-0034-01 17-0032-02
95016-760EA 386.55 USD InStock
95016-760 CA95016-760 95016-762 95016-766 95016-754 CA95016-762L 95016-768 95016-756 CA95016-768 CA95016-754 CA95016-766 95016-758 CA95016-758 CA95016-756L
Sephadex™ G-25 Gel Filtration Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex G-25 is well established gel filtration medium for desalting and buffer exchange in industrial applications.
  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Excellent recovery and minimum sample dilution
  • BioProcess resin supported for industrial applications
Sephadex is a gel filtration resin prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex G-25 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large molecules. Sephadex G-25 is available in 4 different particle sizes (Coarse, Medium, Fine and Superfine). Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Coarse and Medium are preferred for large scale group separations where high flow rates and low operating pressures are required.
Sephadex G-25 is also available in prepacked HiPrep™ desalting columns for fast and convenient desalting.
Ordering information: Supplied in lab packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty