Would you like to log in again?
Sephadex™ G-50 Gel Filtration Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sephadex™
17-0043-01 17-0041-01 17-0041-01 17-0042-01 17-0042-02 17-0043-02 17-0043-01 17-0042-01 17-0043-02 17-0042-02
95016-774EA 1908.16 USD InStock
CA95016-776L 95016-774 CA95016-774L 95016-776 CA95016-772L 95016-778 CA95016-770L 95016-770 CA95016-778L 95016-772
Sephadex™ G-50 Gel Filtration Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex G-50 Medium is well established gel filtration medium for desalting and buffer exchange of biomolecules >30 000 molecular weight.
  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Suitable for purification of DNA from small molecules by gel filtration
  • Larger bead size for large scale group separations where high flow rates and low operating pressures are required
  • Classic gel filtration medium
Sephadex is a gel filtration resin prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-50 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large molecules. Sephadex G-50 is available in 3 different particle sizes (Medium, Fine and Superfine). Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Medium is preferred for large scale group separations where high flow rates and low operating pressures are required.
The larger bead size of G-50 Medium is suited for large scale separations.
Ordering information: Supplied in lab packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty