Would you like to log in again?
DEAE Sephadex™ A-25 Ion Exchange Chromatography Media
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sephadex™
17-0170-01 17-0170-02 17-0170-01 17-0170-02
95055-928EA 572.85 USD InStock
95055-928 95016-802 CA95055-928L CA95016-802L
DEAE Sephadex™ A-25 Ion Exchange Chromatography Media
Protein Purification Protein Purification Resins
DEAE Sephadex™ A-25 and DEAE Sephadex™ A-50 are weak anion exchangers. Sephadex™ ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.
  • Suitable for batch techniques
  • High binding capacities
  • Well-proven and widely used especially in approved plasma fractionation processes (DEAE Sephadex™ A-50 only)
Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty