Would you like to log in again?
DEAE Sephadex™ A-50 Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Sephadex™
17-0180-01 17-0180-02 17-0180-02 17-0180-01
95016-804EA 2541.6 USD InStock
CA95055-930L 95016-804 CA95016-804L 95055-930
DEAE Sephadex™ A-50 Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
DEAE Sephadex™ A-25 and DEAE Sephadex™ A-50 are weak anion exchangers. Sephadex™ ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.
  • Weak anion exchanger suitable for batch techniques
  • High binding capacities
  • Well-proven and widely used
  • Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride
These groups are attached to glucose units in the matrix by stable ether linkages. Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty