Would you like to log in again?
Con A Sepharose™ 4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0440-03 17-0440-01 17-0440-01 17-0440-03
95016-822EA 1201.79 USD InStock
95016-822 95016-824 CA95016-824L CA95016-822L
Con A Sepharose™ 4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Con A Sepharose is Concanavalin A coupled to Sepharose 4B by the cyanogen bromide method.
  • Group-specific adsorbent for molecules containing sugars
  • Con A coupled to Sepharose 4B via cyanogen bromide activation
  • Highly suitable for isolation of cell surface glycoproteins from detergent-solubilized membranes
  • Cyanogen bromide activation coupling method
Concanavalin A (Con A) is a tetrameric metalloprotein isolated from Canavalia ensiformis (jack bean). Con A binds molcules containing α-D-mannopyranosyl, α-D-glucopyranosyl and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups. The binding sugar requires the presence of C-3, C-4 and C-5 hydroxyl groups for reaction with Con A. Con A coupled to Sepharose is routinely used for separation and purification of glycoproteins, polysaccharides and glycolipids. Chelating agents such as EDTA, 8 M urea or solutions having a pH below 3 should be avoided as these conditions results in removal of manganese from the lectin with loss of activity as a result.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty