Would you like to log in again?
Lentil Lectin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0444-01 17-0444-01
95016-830EA 1037.09 USD InStock
95016-830 CA95016-830L
Lentil Lectin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Lentil lectin is a metalloprotein, containing Ca₂₊ and Mn₂₊, which binds reversibly to polysaccharides and glycoconjugates containing glucose or mannose type sugars
  • For affinity chromatography
  • Cyanogen bromide activation coupling methods
  • Stable to all commonly used aqueous buffers
  • Chelating agents should be avoided
  • Negligible volume variation due to changes in pH or ionic strength
Lentil Lectin Sepharose 4B is a generally applicable group specific adsorbent and is routinely used in the preparation and purification of a number of glycoproteins and carbohydrate containing molecules. Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues. Compared to Con A, also for glycoprotein purification, Lentil Lectin distinguishes less sharply between glucosyl and mannosyl. Retains binding in 1% deoxycholate and is especially valuable for purification of detergent-solubilized membrane proteins. Chelating agents such as EDTA, 8 M urea or solutions having a pH below 3 should be avoided as these conditions result in removal of manganese from the lectin and loss of activity
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty