Would you like to log in again?
Epoxy-Activated Sepharose™ 6B Affinity Coupling Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0480-01 17-0480-01
95016-858EA 1005.91 USD InStock
95016-858 CA95016-858L
Epoxy-Activated Sepharose™ 6B Affinity Coupling Media, Cytiva
Protein Purification Protein Purification Resins
Epoxy-activated Sepharose™ 6B is a pre-activated medium for immobilisation of various ligands including sugars through coupling of hydroxy, amino or thiol groups on the ligand to Sepharose™ 6B via a 12-atom hydrophilic spacer arm. Medium is formed by reacting Sepharose™ 6B with 1,4-bis (2,3- epoxy-propoxy-) butane. It can be used to couple sugars and other carbohydrates via stable ether linkages to hydroxyl groups.
Epoxy-activated Sepharose 6B is a pre-activated medium for immobilization of various ligands. Other ligands can be coupled through hydroxyl, amino or thiol groups. The medium has a long hydrophilic spacer arm which makes it particular suitable for immobilization of small molecules. Epoxy-activated Sepharose 6B is formed by reacting Sepharose 6B with 1,4-bis (2,3- epoxy-propoxy-) butane. Stable to all commonly used aqueous solutions. Can be used with non-ionic detergents, denaturing solvents, e.g. 8 M urea and 6 M guanidine hydrochloride. Stable in organic solvents, e.g. 50% dimethylformamide and 50% dioxane. Negligible volume variation due to changes in pH or ionic strength.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty