Would you like to log in again?
Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0529-01 17-0529-01
95016-874EA 701.13 USD InStock
95016-874 CA95016-874L
Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Calmodulin Sepharose™ 4B is calmodulin immobilised by the CNBr method to Sepharose™ 4B. Calmodulin is a highly conserved regulatory protein found in all eukaryotic cells. This protein is involved in many cellular processes such as glycogen metabolism, cytoskeletal control, neurotransmission, phosphate activity and control of NAD⁺/NADP⁺.
  • For single-step purification of native calmodulin-binding proteins
  • Suitable for tandem affinity purification (TAP) of protein complexes
  • Purification of calmodulin-regulated proteins from all eukaryotic cells
  • Cyanogen bromide activation coupling method
  • Stable to all commonly used aqueous solutions
  • Negligible volume variation due to changes in pH or ionic strength
Calmodulin binds proteins principally through their interactions with hydrophobic sites on its surface. These sites are exposed after a conformational change induced by the action of Ca²⁺ on separate Ca²⁺ binding sites. The binding of enzymes may be enhanced if the enzyme substrate is present and enzymesubstratecalmoldulin-Ca²⁺ complexes are particularly stable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty