Would you like to log in again?
Protein G Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0618-02 17-0618-05 17-0618-01 17-0618-01 17-0618-02 17-0618-05
95016-924EA 858.82 USD InStock
CA95016-928L 95016-924 CA95016-926L 95016-926 CA95016-924L 95016-928
Protein G Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Protein G Sepharose 4 Fast Flow has recombinant protein G immobilized by the cyanogen bromide (CNBr) method to Sepharose 4 Fast Flow.
  • For antibody affinity chromatography
  • Fractionation of monoclonal antibodies without binding albumin
  • Recombinant protein G lacking albumin-binding region
  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • Cyanogen bromide activation coupling method
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of research applications
As member of the BioProcess media range, Protein G Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Protein G exhibit binding specificities that complement Protein A media and binds to the Fc region of IgG from a variety of mammalian species. Protein G Sepharose 4 Fast Flow may be used to isolate and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty