Would you like to log in again?
Glutathione Sepharose™ 4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0756-01 17-0756-04 17-0756-05 17-0756-01 17-0756-04 17-0756-05
95016-984EA 391.02 USD InStock
CA95016-984L 95016-984 95016-986 89128-878 CA89128-878 CA95016-986L
Glutathione Sepharose™ 4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Glutathione Sepharose™ 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.
  • Proteins are eluted under mild, non-denaturing conditions that preserve protein antigenicity and functionality
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty